RIE156321267 Rieke Metals, Inc.

4-Bromo-2'-chlorobenzophenone

  • MDL number MFCD09801457
  • Cas Number : 464190-33-8
  • Molecular Weight : 295.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5182-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5182-19
Availability details for
Quantity
  • Availability
  • Add to Cart