RIE156321269 Rieke Metals, Inc.

3-(2-Pyridinyl)cyclohexanone

  • MDL number MFCD03841027
  • Cas Number : 110225-73-5
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5557

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5557
Availability details for
Quantity
  • Availability
  • Add to Cart