RIE156321271 Rieke Metals, Inc.

4-fluoro-4'-nitrobenzophenone

  • MDL number MFCD00053428
  • Cas Number : 2195-47-3
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5100

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5100
Availability details for
Quantity
  • Availability
  • Add to Cart