RIE156321272 Rieke Metals, Inc.

4-cyano-4'-nitrobenzophenone

  • MDL number MFCD03841021
  • Cas Number : 68271-77-2
  • Molecular Weight : 252.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5171

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5171
Availability details for
Quantity
  • Availability
  • Add to Cart