RIE156321273 Rieke Metals, Inc.

3-Benzoylthiophene

  • MDL number MFCD00114995
  • Cas Number : 6453-99-2
  • Molecular Weight : 188.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-01
Availability details for
Quantity
  • Availability
  • Add to Cart