RIE156321278 Rieke Metals, Inc.

4-Chloro-3',4'-dimethylbenzophenone

  • MDL number MFCD06201507
  • Cas Number : 40415-10-9
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5196-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5196-16
Availability details for
Quantity
  • Availability
  • Add to Cart