RIE156321291 Rieke Metals, Inc.

3,3',4'-Trichlorobenzophenone

  • MDL number MFCD06201486
  • Cas Number : 812684-89-2
  • Molecular Weight : 285.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(3,4-dichlorophenyl)methanone
Inchi Code 1S/C13H7Cl3O/c14-10-3-1-2-8(6-10)13(17)9-4-5-11(15)12(16)7-9/h1-7H
Inchi Key UGTVFNDTQOCKNT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5183-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5183-47
Availability details for
Quantity
  • Availability
  • Add to Cart