RIE156321293 Rieke Metals, Inc.

3-Chloro-3',5'-dimethylbenzophenone

  • MDL number MFCD06201492
  • Cas Number : 844884-98-6
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5197-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5197-15
Availability details for
Quantity
  • Availability
  • Add to Cart