RIE156321296 Rieke Metals, Inc.

3-Bromo-3',5'-dimethylbenzophenone

  • MDL number MFCD06201472
  • Cas Number : 844879-51-2
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5197-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5197-18
Availability details for
Quantity
  • Availability
  • Add to Cart