RIE156321299 Rieke Metals, Inc.

4-Chloro-4'-fluoro-3'-methylbenzophenone

  • MDL number MFCD09801644
  • Cas Number : 59612-89-4
  • Molecular Weight : 248.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)(4-fluoro-3-methylphenyl)methanone
Inchi Code 1S/C14H10ClFO/c1-9-8-11(4-7-13(9)16)14(17)10-2-5-12(15)6-3-10/h2-8H,1H3
Inchi Key ORARFWXKHBAQIL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5209-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5209-16
Availability details for
Quantity
  • Availability
  • Add to Cart