RIE156321301 Rieke Metals, Inc.

3-Bromo-4'-fluoro-3'-methylbenzophenone

  • MDL number MFCD06201476
  • Cas Number : 844884-90-8
  • Molecular Weight : 293.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5209-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5209-18
Availability details for
Quantity
  • Availability
  • Add to Cart