RIE156321304 Rieke Metals, Inc.

3,4,4'-Trichlorobenzophenone

  • MDL number MFCD06201503
  • Cas Number : 33093-42-4
  • Molecular Weight : 285.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)(3,4-dichlorophenyl)methanone
Inchi Code 1S/C13H7Cl3O/c14-10-4-1-8(2-5-10)13(17)9-3-6-11(15)12(16)7-9/h1-7H
Inchi Key WGEYEANIOWFMGC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5184-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5184-47
Availability details for
Quantity
  • Availability
  • Add to Cart