RIE156321314 Rieke Metals, Inc.

2-Acetoxybenzophenone

  • MDL number MFCD07698796
  • Cas Number : 138711-39-4
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-benzoylphenyl acetate
Inchi Code 1S/C15H12O3/c1-11(16)18-14-10-6-5-9-13(14)15(17)12-7-3-2-4-8-12/h2-10H,1H3
Inchi Key ZTTANZQQBIQFEG-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-01
Availability details for
Quantity
  • Availability
  • Add to Cart