RIE156321315 Rieke Metals, Inc.

3-Bromo-4'-isopropylbenzophenone

  • MDL number MFCD06201460
  • Cas Number : 844879-27-2
  • Molecular Weight : 303.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5210-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5210-18
Availability details for
Quantity
  • Availability
  • Add to Cart