RIE156321316 Rieke Metals, Inc.

4-Bromo-4'-isopropylbenzophenone

  • MDL number MFCD06201442
  • Cas Number : 91404-24-9
  • Molecular Weight : 303.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5210-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5210-19
Availability details for
Quantity
  • Availability
  • Add to Cart