RIE156321318 Rieke Metals, Inc.

4-Chloro-3',5'-dimethyl-4'-methoxybenzophenone

  • MDL number MFCD06201506
  • Cas Number : 61259-84-5
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)(4-methoxy-3,5-dimethylphenyl)methanone
Inchi Code 1S/C16H15ClO2/c1-10-8-13(9-11(2)16(10)19-3)15(18)12-4-6-14(17)7-5-12/h4-9H,1-3H3
Inchi Key VCOQTPORNRFKDV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5198-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5198-16
Availability details for
Quantity
  • Availability
  • Add to Cart