RIE156321321 Rieke Metals, Inc.

3,4-Difluoro-4'-isopropylbenzophenone

  • MDL number MFCD06201556
  • Cas Number : 845781-00-2
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5210-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5210-41
Availability details for
Quantity
  • Availability
  • Add to Cart