RIE156321325 Rieke Metals, Inc.

Cyclohexyl 3-thienyl ketone

  • MDL number MFCD07699057
  • Cas Number : 36646-69-2
  • Molecular Weight : 194.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-04
Availability details for
Quantity
  • Availability
  • Add to Cart