RIE156321333 Rieke Metals, Inc.

4-n-Butyl-3'-chlorobenzophenone

  • MDL number MFCD06201484
  • Cas Number : 844884-93-1
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5180-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-15
Availability details for
Quantity
  • Availability
  • Add to Cart