RIE156321335 Rieke Metals, Inc.

3,4-Difluoro-3',5'-dimethyl-4'-methoxybenzophenone

  • MDL number MFCD06201565
  • Cas Number : 845781-08-0
  • Molecular Weight : 276.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)(4-methoxy-3,5-dimethylphenyl)methanone
Inchi Code 1S/C16H14F2O2/c1-9-6-12(7-10(2)16(9)20-3)15(19)11-4-5-13(17)14(18)8-11/h4-8H,1-3H3
Inchi Key NTEYQUCAZAUNNI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5198-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5198-41
Availability details for
Quantity
  • Availability
  • Add to Cart