RIE156321337 Rieke Metals, Inc.

3,4-Dichloro-3',5'-dimethyl-4'-methoxybenzophenone

  • MDL number MFCD06201547
  • Cas Number : 61259-86-7
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5198-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5198-47
Availability details for
Quantity
  • Availability
  • Add to Cart