RIE156321338 Rieke Metals, Inc.

4-chloro-3'-nitrobenzophenone

  • MDL number MFCD00617171
  • Cas Number : 62810-38-2
  • Molecular Weight : 261.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)(3-nitrophenyl)methanone
Inchi Code 1S/C13H8ClNO3/c14-11-6-4-9(5-7-11)13(16)10-2-1-3-12(8-10)15(17)18/h1-8H
Inchi Key WANOJUMUCWLVTE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5088

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5088
Availability details for
Quantity
  • Availability
  • Add to Cart