RIE156321340 Rieke Metals, Inc.

3-Bromo-4'-n-Butylbenzophenone

  • MDL number MFCD06201464
  • Cas Number : 844879-33-0
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5180-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-18
Availability details for
Quantity
  • Availability
  • Add to Cart