RIE156321344 Rieke Metals, Inc.

4-Chloro-3'-methylbenzophenone

  • MDL number MFCD06201495
  • Cas Number : 35256-82-7
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5212-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5212-16
Availability details for
Quantity
  • Availability
  • Add to Cart