RIE156321349 Rieke Metals, Inc.

4-Bromo-4'-chlorobenzophenone

  • MDL number MFCD00045208
  • Cas Number : 27428-57-5
  • Molecular Weight : 295.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5161

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5161
Availability details for
Quantity
  • Availability
  • Add to Cart