RIE156321350 Rieke Metals, Inc.

4-n-Butyl-3',4'-difluorobenzophenone

  • MDL number MFCD06201560
  • Cas Number : 845781-04-6
  • Molecular Weight : 274.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butylphenyl)(3,4-difluorophenyl)methanone
Inchi Code 1S/C17H16F2O/c1-2-3-4-12-5-7-13(8-6-12)17(20)14-9-10-15(18)16(19)11-14/h5-11H,2-4H2,1H3
Inchi Key PRQKSSOAHRWKSW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5180-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-41
Availability details for
Quantity
  • Availability
  • Add to Cart