RIE156321351 Rieke Metals, Inc.

4-tert-Butyl-3'-methylbenzophenone

  • MDL number MFCD09801672
  • Cas Number : 55709-37-0
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5212-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5212-34
Availability details for
Quantity
  • Availability
  • Add to Cart