RIE156321353 Rieke Metals, Inc.

3,5-Difluoro-3'-methylbenzophenone

  • MDL number MFCD06201511
  • Cas Number : 844885-06-9
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5212-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5212-42
Availability details for
Quantity
  • Availability
  • Add to Cart