RIE156321354 Rieke Metals, Inc.

3,4-Dichloro-3'-methylbenzophenone

  • MDL number MFCD06201532
  • Cas Number : 844885-24-1
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dichlorophenyl)(3-methylphenyl)methanone
Inchi Code 1S/C14H10Cl2O/c1-9-3-2-4-10(7-9)14(17)11-5-6-12(15)13(16)8-11/h2-8H,1H3
Inchi Key OJGLIPKGQBRCAH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5212-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5212-47
Availability details for
Quantity
  • Availability
  • Add to Cart