RIE156321356 Rieke Metals, Inc.

2-Chloro-4'-methylbenzophenone

  • MDL number MFCD01463397
  • Cas Number : 5953-00-4
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-14
Availability details for
Quantity
  • Availability
  • Add to Cart