RIE156321357 Rieke Metals, Inc.

3-Chloro-4'-methylbenzophenone

  • MDL number MFCD01662298
  • Cas Number : 13395-60-3
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-15
Availability details for
Quantity
  • Availability
  • Add to Cart