RIE156321358 Rieke Metals, Inc.

4-Chloro-4'-methylbenzophenone

  • MDL number MFCD00625699
  • Cas Number : 5395-79-9
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-16
Availability details for
Quantity
  • Availability
  • Add to Cart