RIE156321361 Rieke Metals, Inc.

4-Bromo-4'-methylbenzophenone

  • MDL number MFCD00625706
  • Cas Number : 76693-57-7
  • Molecular Weight : 275.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-19
Availability details for
Quantity
  • Availability
  • Add to Cart