RIE156321367 Rieke Metals, Inc.

3,4-Dichloro-4'-methylbenzophenone

  • MDL number MFCD06201533
  • Cas Number : 125016-15-1
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-47
Availability details for
Quantity
  • Availability
  • Add to Cart