RIE156321373 Rieke Metals, Inc.

3,3',4,5'-Tetrachlorobenzophenone

  • MDL number MFCD06201544
  • Cas Number : 844885-32-1
  • Molecular Weight : 320.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5190-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5190-48
Availability details for
Quantity
  • Availability
  • Add to Cart