RIE156321374 Rieke Metals, Inc.

4-n-Butyl-3',4'-dichlorobenzophenone

  • MDL number MFCD06201541
  • Cas Number : 844885-30-9
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5180-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-47
Availability details for
Quantity
  • Availability
  • Add to Cart