RIE156321378 Rieke Metals, Inc.

4-Bromo-3'-chloro-5'-fluorobenzophenone

  • MDL number MFCD06201449
  • Cas Number : 844879-12-5
  • Molecular Weight : 313.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5186-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-19
Availability details for
Quantity
  • Availability
  • Add to Cart