RIE156321379 Rieke Metals, Inc.

4-ethoxy-2',3',4',5',6'-pentafluorobenzophenone

  • MDL number MFCD03427112
  • Cas Number : 351003-31-1
  • Molecular Weight : 316.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethoxyphenyl)(2,3,4,5,6-pentafluorophenyl)methanone
Inchi Code 1S/C15H9F5O2/c1-2-22-8-5-3-7(4-6-8)15(21)9-10(16)12(18)14(20)13(19)11(9)17/h3-6H,2H2,1H3
Inchi Key YZUXMWWGIFJHOY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5162

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5162
Availability details for
Quantity
  • Availability
  • Add to Cart