RIE156321380 Rieke Metals, Inc.

3,5-Dichloro-3',4'-difluorobenzophenone

  • MDL number MFCD06201562
  • Cas Number : 845781-05-7
  • Molecular Weight : 287.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5191-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5191-41
Availability details for
Quantity
  • Availability
  • Add to Cart