RIE156321381 Rieke Metals, Inc.

3,5-Dichloro-3',5'-difluorobenzophenone

  • MDL number MFCD06201525
  • Cas Number : 844885-17-2
  • Molecular Weight : 287.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5191-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5191-42
Availability details for
Quantity
  • Availability
  • Add to Cart