RIE156321384 Rieke Metals, Inc.

3-Chloro-4'-n-propylbenzophenone

  • MDL number MFCD06201483
  • Cas Number : 64358-13-0
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(4-propylphenyl)methanone
Inchi Code 1S/C16H15ClO/c1-2-4-12-7-9-13(10-8-12)16(18)14-5-3-6-15(17)11-14/h3,5-11H,2,4H2,1H3
Inchi Key YSSHSXYMACHHFY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5215-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-15
Availability details for
Quantity
  • Availability
  • Add to Cart