RIE156321393 Rieke Metals, Inc.

3,4-Difluoro-4'-n-propylbenzophenone

  • MDL number MFCD06201559
  • Cas Number : 845781-03-5
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5215-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-41
Availability details for
Quantity
  • Availability
  • Add to Cart