RIE156321394 Rieke Metals, Inc.

2-(4-Chlororobenzoyl)-5-(1,3-Dioxolan-2-yl)thiophene

  • MDL number MFCD07699132
  • Cas Number : 898773-50-7
  • Molecular Weight : 294.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)[5-(1,3-dioxolan-2-yl)-2-thienyl]methanone
Inchi Code 1S/C14H11ClO3S/c15-10-3-1-9(2-4-10)13(16)11-5-6-12(19-11)14-17-7-8-18-14/h1-6,14H,7-8H2
Inchi Key IYIHKIZLLVKAFX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5568-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5568-16
Availability details for
Quantity
  • Availability
  • Add to Cart