RIE156321396 Rieke Metals, Inc.

3,4-Dichloro-4'-n-propylbenzophenone

  • MDL number MFCD06201540
  • Cas Number : 844885-29-6
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5215-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-47
Availability details for
Quantity
  • Availability
  • Add to Cart