RIE156321400 Rieke Metals, Inc.

3-Chloro-3',5,5'-trifluorobenzophenone

  • MDL number MFCD06201526
  • Cas Number : 844885-18-3
  • Molecular Weight : 270.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-5-fluorophenyl)(3,5-difluorophenyl)methanone
Inchi Code 1S/C13H6ClF3O/c14-9-1-7(2-10(15)5-9)13(18)8-3-11(16)6-12(17)4-8/h1-6H
Inchi Key XFGRJWZHVBYIPC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5186-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-42
Availability details for
Quantity
  • Availability
  • Add to Cart