RIE156321402 Rieke Metals, Inc.

3,4-Difluoro-4'-ethylbenzophenone

  • MDL number MFCD06201558
  • Cas Number : 845781-02-4
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5201-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5201-41
Availability details for
Quantity
  • Availability
  • Add to Cart