RIE156321407 Rieke Metals, Inc.

3-Chloro-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD06014620
  • Cas Number : 727994-89-0
  • Molecular Weight : 274.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-15
Availability details for
Quantity
  • Availability
  • Add to Cart