RIE156321408 Rieke Metals, Inc.

4-Chloro-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD01105757
  • Cas Number : 101018-98-8
  • Molecular Weight : 274.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-16
Availability details for
Quantity
  • Availability
  • Add to Cart