RIE156321410 Rieke Metals, Inc.

3-Chloro-4'-(thiomethyl)benzophenone

  • MDL number MFCD06201479
  • Cas Number : 197438-99-6
  • Molecular Weight : 262.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)[4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C14H11ClOS/c1-17-13-7-5-10(6-8-13)14(16)11-3-2-4-12(15)9-11/h2-9H,1H3
Inchi Key LQNJUYUSDLKVQG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5217-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-15
Availability details for
Quantity
  • Availability
  • Add to Cart