RIE156321412 Rieke Metals, Inc.

3-Bromo-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD05858892
  • Cas Number : 727419-55-8
  • Molecular Weight : 319.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-18
Availability details for
Quantity
  • Availability
  • Add to Cart